N-(3-Aminopropyl)-N-methylaniline

Supplier: SPECTRUM CHEMICAL MFG CORP

A1484-100ML A1484-25ML
75871-710EA 396.2 USD
75871-710 75871-712
N-(3-Aminopropyl)-N-methylaniline
N-(3-Aminopropyl)-N-methylaniline
Formula: C₁₀H₁₆N₂
MW: 164.250 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191602
CAS Number: 53485-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR