Papanicolaou's solution EA 36, HARLECO® stain for cytology, Sigma-Aldrich®
Supplier: MilliporeSigma
7058X-85 7058X-75
15204-212EA 228 USD
15204-212 15204-270
Papanicolaou's solution EA 36, HARLECO® stain for cytology, Sigma-Aldrich®
Papanicolaou's solution EA 36
Cytoplasmic counterstain for Pap staining technique.
Prepared from dyes certified by the Biological Stain Commission (BSC)