Giemsa's stain, Technical Grade used as a blood stain, Sigma-Aldrich®

Warning

Synonyms: Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue

G4507-5G G4507-25G
220179-278EA 142.8 USD
220179-278 220179-277
Giemsa's stain, Technical Grade used as a blood stain, Sigma-Aldrich®
Giemsa's stain
Storage Temperature: Ambient
MDL Number: MFCD00081642
CAS Number: 51811-82-6
EINECS: 257-438-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65a3dde4b74ec23e6f527c0ce338d61d470c8d26&vid=c7baef9be683f2300682a16b9432418e0a4311a0&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR