Giemsa's stain modified solution, Sigma-Aldrich®

Danger

Synonyms: Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue

32884-1L 32884-250ML
220056-360EA 161 USD
220056-360 220056-361
Giemsa's stain modified solution, Sigma-Aldrich®
Giemsa's stain

Modified Giemsa stain may be used for the identification of each human chromosome by differentials staining.


Modified Giemsa stain means when the pH of the Giemsa stain is changed from the usual 6.8 to 9.0.

Density: 0.99 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00081642
CAS Number: 51811-82-6
EINECS: 257-438-2
UN: 1230
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5d57f471bcd5db3202c766f3a3ba16950cfc499&vid=c5d57f471bcd5db3202c766f3a3ba16950cfc499&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22b87821d8c140943a6f068261320ff57f4bb55a&vid=22b87821d8c140943a6f068261320ff57f4bb55a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR