(2-Methoxyethoxy)acetic acid ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 2-(2-Methoxyethoxy)acetic acid

143402-1G 143402-10G
103627-826EA 39.1 USD
103627-826 103627-828
(2-Methoxyethoxy)acetic acid ≥97%
(2-Methoxyethoxy)acetic acid
Formula: C₅H₁₀O₄
MW: 134.13 g/mol
MDL Number: MFCD00044097
CAS Number: 16024-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR