3,5-Dibromobenzaldehyde ≥99% (by GC)

Supplier: CHEM IMPEX INTL INC MS

28106-25G 28106-1G 28106-5G
102570-642EA 164.68 USD
102570-642 102605-140 102610-646
3,5-Dibromobenzaldehyde ≥99% (by GC)
3,5-Dibromobenzaldehyde
Formula: C₇H₄Br₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00156887
CAS Number: 56990-02-4

Order Now

Specification Test Results

Appearance Off-White to Light Yellow Solid
Water Content (Karl Fisher) ≤0.5%
Residual Solvents ≤0.5%
¹H NMR Conforms to Structure
Purity by GC ≥99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR