R-Phycoerythrin ≥10.0 mg/mL (UV), BioReagent

52412-5MG-F 52412-1MG-F
220074-996EA 1021.25 USD
220074-996 220074-995
R-Phycoerythrin ≥10.0 mg/mL (UV), BioReagent
R-Phycoerythrin

-Phycoerythrin (R-PE), is useful in the laboratory as a fluorescence-based indicator for the presence of cyanobacteria and for a variety of immunofluorescence applications. R-PE is a useful fluoroprobe in various electrophoretic procedures.


  • fluorescence : λex 542 nm; λem 575 nm in 0.1 M phosphate pH 7.2
  • suitability : passes test for gel electrophoresis

MW: 240 kDa
Storage Temperature: Refrigerator
MDL Number: MFCD00081950
CAS Number: 11016-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR