7-Amino-4-methyl-3-coumarinylacetic acid (AMCA-H) ~90% (by HPLC), BioReagent fluorescent dye, Sigma-Aldrich®

Warning

Synonyms: AMCA, 2H-1-Benzopyran-3-acetic acid, 7-Amino-4-methylcoumarin-3-acetic acid, Aminomethyl coumarin acetic acid, 2-(7-Amino-4-methyl-2-oxo-2H-chromen-3-yl)acetic acid

08445-100MG 08445-500MG
220032-778EA 135.18 USD
220032-778 220032-779
7-Amino-4-methyl-3-coumarinylacetic acid (AMCA-H) ~90% (by HPLC), BioReagent fluorescent dye, Sigma-Aldrich®
7-Amino-4-methyl-3-coumarinylacetic acid (AMCA-H)

AMCA is a useful agent for labeling proteins. Demonstrates a characteristic absorption peak at 345 nm with a fluorescence emission at 440-460 nm (blue region); however, when conjugated with proteins the absorption peak shifts to 355 nm with a fluorescence emission at 440-460 nm. It has a high fluorescence yield with a sufficient Stokes shift (100 nm) and is relatively photostable. Relatively easy to activate and couple to the N-terminal amino group of a peptide resulting in an acid stable amide bond.


  • Fluorescence - λex 350 nm; λem 433 nm in methanol

Formula: C₁₂H₁₁NO₄
MW: 233.22 g/mol
MDL Number: MFCD00081051
CAS Number: 106562-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR