Zinc bromide, anhydrous ≥98%, Puriss., Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Zinc (II) bromide

02128-1KG 02128-4X5KG
BJ02128-1KGEA 502 USD
BJ02128-1KG BJ02128-500G BJ02128-5KG
Zinc bromide, anhydrous ≥98%, Puriss., Fluka™
Zinc bromide
Formula: Br₂Zn
MW: 225.2 g/mol
Boiling Pt: 697 °C (1.013 hPa)
Melting Pt: 394
Density: 4.22 g/cm³ (20 °C)
Storage Temperature: Room Temperature
MDL Number: MFCD00011294
CAS Number: 7699-45-8
EINECS: 231-718-4
UN: 3260
ADR: 3 III
Merck Index: 13,10182

Order Now

Specification Test Results

Assay Min. 98 %
Barium (Ba) Max. 0.001 %
Calcium (Ca) Max. 0.01 %
Iron (Fe) Max. 0.001 %
Potassium (K) Max. 0.01 %
Magnesium (Mg) Max. 0.01 %
Sodium (Na) Max. 0.01 %
Lead (Pb) Max. 0.005 %
Sulfate (SO₄) Max. 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR