Triethylammonium phosphate solution_SC ~1.0 M water, BioUltra, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: TEAP

90361-100ML 90361-500ML
BJ90361-100MLEA 153 USD
BJ90361-100ML BJ90361-500ML
Triethylammonium phosphate solution_SC ~1.0 M water, BioUltra, Fluka™
Triethylammonium phosphate
Formula: C₆H₁₈NO₄P
MW: 199.19 g/mol
Density: 1.050 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00067475
CAS Number: 10138-93-9

Order Now

Specification Test Results

Assay 0.9 - 1.1 mol/l
Refractive index (n 20/D) 1.358 - 1.362
pH (direct, 25 °C) 2.9 - 3.1
Arsenic (As) Max. 0.1 ppm
Aluminium (Al) Max. 1 ppm
Barium (Ba) Max. 1 ppm
Bismuth (Bi) Max. 1 ppm
Calcium (Ca) Max. 5 ppm
Cadmium (Cd) Max. 1 ppm
Cobalt (Co) Max. 1 ppm
Chromium (Cr) Max. 1 ppm
Copper (Cu) Max. 1 ppm
Iron (Fe) Max. 1 ppm
Potassium (K) Max. 20 ppm
Lithium (Li) Max. 1 ppm
Magnesium (Mg) Max. 1 ppm
Manganese (Mn) Max. 1 ppm
Molybdenum (Mo) Max. 1 ppm
Sodium (Na) Max. 20 ppm
Nickel (Ni) Max. 1 ppm
Lead (Pb) Max. 1 ppm
Strontium (Sr) Max. 1 ppm
Zinc (Zn) Max. 1 ppm
Sulfate (SO₄) Max. 10 ppm
Chloride (Cl) Max. 10 ppm
Filter test complying
Absorbance 280 nm Max. 0.03
Absorbance at 260 nm Max. 0.04


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR