Sodium tungstate dihydrate ≥99% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Tungstic acid sodium salt dihydrate, Sodium wolframate dihydrate, Sodium tungsten oxide dihydrate

223336-100G 223336-500G
BJ223336-100GEA 246 USD
BJ223336-100G BJ223336-500G
Sodium tungstate dihydrate ≥99% ACS, Fluka™
Sodium tungstate dihydrate
Formula: Na₂WO₄·2H₂O
MW: 329.85 g/mol
Melting Pt: 698 °C
Density: 4.18 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149190
CAS Number: 10213-10-2

Order Now

Specification Test Results

Appearance complying
Assay 99.0 - 101.0 %
Titrable Base Max. 0.02 meq/g
Water insoluble matter Max. 0.01 %
Molybdenum (Mo) Max. 0.001 %
Chloride (Cl) Max. 0.005 %
Heavy metals (as Pb) and iron Max. 0.001 %
Sulfate (SO₄) Max. 0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR