Sodium sulfate, anhydrous ≥99.0%, powder ACS, Fluka™
Supplier: Honeywell Research Chemicals
238597-1KG
238597-2.5KG
238597-500G
BJ238597-1KGEA
79
USD
BJ238597-1KG
BJ238597-2.5KG
BJ238597-500G
Sodium sulfate, anhydrous ≥99.0%, powder ACS, Fluka™
Sodium sulphate
Formula:
Na₂SO₄ Melting Pt: 884 °C Density: 2.68 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003504 CAS Number: 7757-82-6 EINECS: 231-820-9 |
|
Specification Test Results
Identity | complying |
Assay | Min. 99.0 % |
pH (5 %, 25 °C) | 5.2 - 9.2 |
Loss on ignition (800°C) | Max. 0.5 % |
Water insoluble matter | Max. 0.01 % |
Calcium (Ca) | Max. 0.01 % |
Iron (Fe) | Max. 0.001 % |
Potassium (K) | Max. 0.01 % |
Magnesium (Mg) | Max. 0.005 % |
Heavy metals (as Pb) | Max. 5 ppm |
Chloride (Cl) | Max. 0.001 % |
Phosphate (PO₄) | Max. 0.001 % |
Total N | Max. 5 ppm |
pH Range | 5.2 - 9.2 |
Form | powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...