Sodium nitroprusside dihydrate ≥99%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Sodium pentacyanonitrosylferrate(III) dihydrate, Sodium nitrosopentacyanoferrate (III) dihydrate, Sodium nitroferricyanide(III) dihydrate, Disodium pentacyanonitrosylferrate (III) dihydrate, Sodium nitroferricyanide dihydrate

71778-100G 71778-25G
BJ71778-100GEA 263.73 USD
BJ71778-100G BJ71778-25G
Sodium nitroprusside dihydrate ≥99%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Sodium nitroprusside dihydrate
Formula: Na₂[Fe(CN)₅NO]·2H₂O
MW: 297.95 g/mol
Density: 1.72 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149192
CAS Number: 13755-38-9
UN: 3288
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Water insoluble matter Max. 0.01 %
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 50 ppm
Copper (Cu) Max. 5 ppm
Potassium (K) Max. 3000 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 50 ppm
Chloride (Cl) Max. 100 ppm
Sulfate (SO₄) Max. 100 ppm
Hexacyanoferrate(II) ([Fe(CN)₆]₄⁻) Max. 200 ppm
Hexacyanoferrate(III) ([Fe(CN)₆]₃⁻) Max. 100 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR