Silicon dioxide for cleaning of platinum crucibles, calcinated, crude, Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
18648-6X2.5KG
18648-50KG
BJ18648-2.5KGCS
739.38
USD
BJ18648-2.5KG
BJ18648-50KG
Silicon dioxide for cleaning of platinum crucibles, calcinated, crude, Fluka™
Silicon dioxide
Formula:
SiO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 7631-86-9 EINECS: 238-878-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...