Sand, quartz, white powder, Purum p.a. >230 mesh, Fluka™

Supplier: Honeywell Research Chemicals
Danger

83340-1KG 83340-5KG
BJ83340-1KGEA 149 USD
BJ83340-1KG BJ83340-5KG
Sand, quartz, white powder, Purum p.a. >230 mesh, Fluka™
Sand, quartz

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2,210 °C(1,013 hPa)
Melting Pt: 1,710 °C
Density: 2.650 g/cm³ (20 °C)
Storage Temperature: Room Temperature
MDL Number: MFCD02100519
CAS Number: 14808-60-7
EINECS: 238-878-4

Order Now

Specification Test Results

Loss on ignition (900°C) Max. 0.3 %
Calcium (Ca) Max. 200 ppm
Cadmium (Cd) Max. 50 ppm
Cobalt (Co) Max. 50 ppm
Copper (Cu) Max. 50 ppm
Iron (Fe) Max. 200 ppm
Potassium (K) Max. 500 ppm
Sodium (Na) Max. 100 ppm
Nickel (Ni) Max. 50 ppm
Lead (Pb) Max. 50 ppm
Zinc (Zn) Max. 50 ppm
Chloride (Cl) Max. 50 ppm
Form powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR