Sand, quartz 50-70 mesh, Fluka™

Supplier: Honeywell Research Chemicals
Danger

274739-1KG 274739-500G 274739-5KG
BJ274739-1KGEA 100 USD
BJ274739-1KG BJ274739-500G BJ274739-5KG
Sand, quartz 50-70 mesh, Fluka™
Sand, quartz
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2.210 °C (1.013 hPa)
Melting Pt: 1.710 °C
Density: 2.650 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD02100519
CAS Number: 14808-60-7
EINECS: 238-878-4

Order Now

Specification Test Results

Particle Size 50-70 mesh
Appearance complying
Granulation 0.21-0.3 mm Min. 85 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR