Potassium iodide ≥99.0% ACS, Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
221945-100G
221945-12KG
221945-2.5KG
221945-500G
BJ221945-100GEA
116.38
USD
BJ221945-100G
BJ221945-12KG
BJ221945-2.5KG
BJ221945-500G
Potassium iodide ≥99.0% ACS, Fluka™
Potassium iodide
Formula:
Kl Boiling Pt: 1.323 °C (1.013 hPa) Melting Pt: 681 °C Density: 3.12 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011405 CAS Number: 7681-11-0 EINECS: 231-659-4 |
Specification Test Results
Assay | Min. 99.0 % |
pH (5 %, 25 °C) | 6.0 - 9.2 |
Loss on drying | Max. 0.2 % |
Water insoluble matter | Max. 0.005 % |
Barium (Ba) | Max. 0.002 % |
Calcium (Ca) | Max. 0.002 % |
Iron (Fe) | Max. 3 ppm |
Magnesium (Mg) | Max. 0.001 % |
Sodium (Na) | Max. 0.005 % |
Heavy metals (as Pb) | Max. 5 ppm |
Bromide, chloride (as Cl) | Max. 0.01 % |
Iodate (IO₃) | Max. 3 ppm |
Phosphate (PO₄) | Max. 0.001 % |
Sulfate (SO₄) | Max. 0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...