Potassium dihydrogen phosphate ≥99.0% ACS, Fluka™

Supplier: Honeywell Research Chemicals

P0662-1KG P0662-2.5KG P0662-500G P0662-6X500G P0662-4X2.5KG
BJP0662-1KGEA 212.22 USD
BJP0662-1KG BJP0662-2.5KG BJP0662-500G BJP0662-6X500G BJP06624X2.5
Potassium dihydrogen phosphate ≥99.0% ACS, Fluka™
Potassium dihydrogen phosphate
Formula: KH₂PO₄
Boiling Pt: Decomposes on heating
Melting Pt: 253 °C
Density: 2.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS Number: 7778-77-0
EINECS: 231-913-4

Order Now

Specification Test Results

Assay Min. 99.0 %
Loss on drying (105°C, 2 h) Max. 0.2 %
pH (5 %, 25 °C) 4.1 - 4.5
Water insoluble matter Max. 0.01 %
Iron (Fe) Max. 0.002 %
Sodium (Na) Max. 0.005 %
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 0.001 %
Sulfate (SO₄) Max. 0.003 %
pKa (25 °C) (1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR