Potassium carbonate ≥98%, powder -325 mesh, Reagent Grade

Supplier: Honeywell Research Chemicals
Warning

347825-1KG 347825-2.5KG
BJ347825-1KGEA 165.34 USD
BJ347825-1KG BJ347825-2.5KG BJ347825-250G
Potassium carbonate ≥98%, powder -325 mesh, Reagent Grade
Potassium carbonate
Formula: K₂CO₃
Boiling Pt: Decomposition
Melting Pt: 891 °C
Density: 2.430 g/cm³ (19 °C)
Storage Temperature: Room Temperature
MDL Number: MFCD00011382
CAS Number: 584-08-7
EINECS: 209-529-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Form powder
Assay 98.0 - 102.0 %
Particle size distribution 50 % Max. 44 µm
Particle Size −325 mesh


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR