Potassium acetate ≥99.0% ACS, Fluka™
Supplier: Honeywell Research Chemicals
Synonyms:
Acetic acid potassium salt
236497-100G
236497-500G
BJ236497-100GEA
50.58
USD
BJ236497-100G
BJ236497-2.5KG
BJ236497-500G
Potassium acetate ≥99.0% ACS, Fluka™
Potassium acetate
Formula:
CH₃COOK MW: 98.14 g/mol Melting Pt: 292 °C Density: 1.57 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00012458 CAS Number: 127-08-2 EINECS: 204-822-2 |
Specification Test Results
Assay (titration with Perchloric acid) | Min. 99.0 % |
Water insoluble matter | Max. 0.005 % |
pH (5 %, 25 °C) | 6.5 - 9.0 |
Calcium (Ca) | Max. 0.005 % |
Iron (Fe) | Max. 5 ppm |
Magnesium (Mg) | Max. 0.002 % |
Sodium (Na) | Max. 0.03 % |
Heavy metals (as Pb) | Max. 5 ppm |
Chloride (Cl) | Max. 0.003 % |
Phosphate (PO₄) | Max. 0.001 % |
Sulfate (SO₄) | Max. 0.002 % |
IR spectrum | complying |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...