o-Cresolphthalein complexone indicator for complexometry, Fluka™
Supplier: Honeywell Research Chemicals
Synonyms:
o-Cresolphthalein Complexone, Phthalein purple, Cresol phthalexone, Xylenolphthalein-bis(iminodiacetic acid), Phthalein complexone, Phthalate complexing agent [Chelation indicator], Metal phthaleine, o-Cresolphthalexon
64000-25G
64000-5G
BJ64000-1GEA
35.78
USD
BJ64000-1G
BJ64000-25G
BJ64000-5G
o-Cresolphthalein complexone indicator for complexometry, Fluka™
o-Cresolphthalein complexone
Formula:
C₃₂H₃₂N₂O₁₂ MW: 636.62 g/mol Melting Pt: 181…185 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00005911 CAS Number: 2411-89-4 EINECS: 219-318-8 |
Specification Test Results
Ignition Residue | ~1% |
Appearance of the substance | complying |
Identity (LC-MS) | complying |
Indicator solution | complying |
Water (Karl Fischer) | Max. 10 % |
Residue on ignition | Max. 2 % |
Suitability for Complexometry | complying |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...