Molybdenum(VI) oxide ≥99.5%, Puriss. p.a., Fluka™
Supplier: Honeywell Research Chemicals
|
Warning
|
69850-100G
69850-500G
BJ69850-100GEA
143.75
USD
BJ69850-100G
BJ69850-500G
Molybdenum(VI) oxide ≥99.5%, Puriss. p.a., Fluka™
Molybdenum(VI) oxide
Formula:
MoO₃ Boiling Pt: 1.155 °C Melting Pt: 795 °C Density: 4.70 g/cm³ (21 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003469 CAS Number: 1313-27-5 EINECS: 215-204-7 |
Specification Test Results
Assay | Min. 99.8 % |
Colour of the solution | complying |
Turbidity of solution | Max. 5.5 NTU |
Calcium (Ca) | Max. 20 ppm |
Cadmium (Cd) | Max. 10 ppm |
Cobalt (Co) | Max. 20 ppm |
Chromium (Cr) | Max. 5 ppm |
Copper (Cu) | Max. 5 ppm |
Iron (Fe) | Max. 20 ppm |
Potassium (K) | Max. 50 ppm |
Magnesium (Mg) | Max. 5 ppm |
Manganese (Mn) | Max. 10 ppm |
Sodium (Na) | Max. 50 ppm |
Nickel (Ni) | Max. 20 ppm |
Lead (Pb) | Max. 20 ppm |
Zinc (Zn) | Max. 20 ppm |
Chloride (Cl) | Max. 20 ppm |
Sulfate (SO₄) | Max. 50 ppm |
Ammonium (NH₄) | Max. 50 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...