Molecular sieve A5 (0.5 nm, 5 Å), beads 8-12 mesh, Fluka™
Supplier: Honeywell Research Chemicals
Synonyms:
Linde 5A, Molecular sieves, 5A 3.0mm-5.0mm pellets
208620-1KG
208620-5KG
BJ208620-1KGEA
223
USD
BJ208620-1KG
BJ208620-5KG
Molecular sieve A5 (0.5 nm, 5 Å), beads 8-12 mesh, Fluka™
Molecular sieve A5 (0.5 nm, 5 Å)
MDL Number:
MFCD00131613 CAS Number: 69912-79-4 |
Specification Test Results
Form | beads |
Grain size 8 - 12 mesh | Min. 95 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...