Molecular sieve 13X (1.0 nm, 10 Å), powder ∼2 μm, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Linde 13X

283592-1KG 283592-5KG
BJ283592-1KGEA 177 USD
BJ283592-1KG BJ283592-5KG
Molecular sieve 13X (1.0 nm, 10 Å), powder ∼2 μm, Fluka™
Molecular sieve 13X (1.0 nm, 10 Å)
Storage Temperature: Ambient
MDL Number: MFCD00164506
CAS Number: 63231-69-6

Order Now

Specification Test Results

Loss on ignition (575°C) Max. 1 %
Water absorbing capacity Min. 20 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR