meta-Phosphoric acid ~100%, lumps (glassy), Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Metaphosphoric acid

04103-10KG 04103-1KG 04103-250G
BJ04103-10KGEA 2810.72 USD
BJ04103-10KG BJ04103-1KG BJ04103-250G
meta-Phosphoric acid ~100%, lumps (glassy), Fluka™
meta-Phosphoric acid
Formula: (HPO₃)n
MW: 79.98 g/mol
Boiling Pt: 600 °C (1.013 hPa)
Melting Pt: 200 °C
Density: 2.0 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011351
CAS Number: 37267-86-0
EINECS: 253-433-4
UN: 3260
ADR: 8,II

Order Now

Specification Test Results

Assay of (HPO₃)n 40 - 50 %
Assay of (naPO₃)n 50 - 60 %
Arsenic (As) Max. 0.0005 %
Iron (Fe) Max. 0.01 %
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR