Mercury(II) iodide ≥99.0% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Mercury diiodide, Mercuric iodide

221090-100G 221090-25G 221090-500G
BJ221090-100GEA 243.26 USD
BJ221090-100G BJ221090-25G BJ221090-500G
Mercury(II) iodide ≥99.0% ACS, Fluka™
Mercury(II) iodide
Formula: HgI₂
MW: 454.4 g/mol
Boiling Pt: 354
Melting Pt: 259
Density: 6.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011044
CAS Number: 7774-29-0
EINECS: 231-873-8
UN: 1638
ADR: 6.1 II
Merck Index: 13,05905

Order Now

Specification Test Results

Assay (excluding dried substances) Min. 99.0 %
Solubility complying
Mercury (Hg⁺) Max. 0.1 %
Soluble mercury salts (as Hg) Max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR