Manganese(IV) oxide ≥90% (RT), Technical Grade, activated, Fluka™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Manganese dioxide, Manganese(IV) oxide, activated, Manganese peroxide, Mangan(IV)-oxid

63548-1KG 63548-250G
BJ63548-1KGEA 354 USD
BJ63548-1KG BJ63548-250G
Manganese(IV) oxide ≥90% (RT), Technical Grade, activated, Fluka™
Manganese(IV) oxide

  • Chemicals that do not have an established standard set for quality and impurity levels or for products where the purity is <90%
  • Variation in color and physical form is possible

Formula: MnO₂
MW: 86.94 g/mol
Melting Pt: Dec.
Density: 5.21 g/cm³ (21 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003463
CAS Number: 1313-13-9
EINECS: 215-202-6

Order Now

Specification Test Results

Appearance complying
Identity complying
Assay Min. 90.0 %
Loss on drying (105°C) Max. 1.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR