Magnesium sulfate hydrate, dried 99.0-101.0% (dried basis), Puriss., meets analytical specification of DAC, Fluka™

Supplier: Honeywell Research Chemicals

13143-6X2.5KG 13143-6X1KG
BJ13143-1KGCS 367.31 USD
BJ13143-1KG BJ13143-2.5KG BJ13143-6X1KG
Magnesium sulfate hydrate, dried 99.0-101.0% (dried basis), Puriss., meets analytical specification of DAC, Fluka™
Magnesium sulphate hydrate
Formula: MgSO₄·nH₂O
Melting Pt: 1.124 °C
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 22189-08-8
EINECS: 231-298-2

Order Now

Specification Test Results

Assay (calculated to the dried substance) 99.0 - 101.0 %
Loss on drying (400°C) 26.0 - 32.0 %
Arsenic (As) Max. 2 ppm
Iron (Fe) Max. 10 ppm
Heavy metals (as Pb) Max. 15 ppm
Chloride (Cl) Max. 400 ppm
Acidity or alkalinity complying
Appearance of the solution complying
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR