Magnesium nitrate hexahydrate 99% ACS, Fluka™
Supplier: Honeywell Research Chemicals
237175-1KG
237175-500G
BJ237175-100GEA
44.14
USD
BJ237175-100G
BJ237175-1KG
BJ237175-2.5KG
BJ237175-500G
Magnesium nitrate hexahydrate 99% ACS, Fluka™
Magnesium nitrate hexahydrate
Formula:
Mg(NO₃)₂·6H₂O MW: 256.41 g/mol Melting Pt: 89 °C Density: 1.464 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149779 CAS Number: 13446-18-9 |
Specification Test Results
Assay | 98.0 - 102.0 % |
pH (5 %, 25 °C) | 5.0 - 8.2 |
Water insoluble matter | Max. 0.005 % |
Barium (Ba) | Max. 0.005 % |
Calcium (Ca) | Max. 0.01 % |
Iron (Fe) | Max. 5 ppm |
Potassium (K) | Max. 0.005 % |
Manganese (Mn) | Max. 5 ppm |
Sodium (Na) | Max. 0.005 % |
Strontium (Sr) | Max. 0.005 % |
Heavy metals (ICP) ACS | Max. 5 ppm |
Chloride (Cl) | Max. 0.001 % |
Phosphate (PO₄) | Max. 5 ppm |
Sulfate (SO₄) | Max. 0.005 % |
Ammonium (NH₄) | Max. 0.003 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...