Lithium carbonate ≥99.0% (by titrimetric analysis), Puriss. p.a. ACS for microscopy, Fluka™

Supplier: Honeywell Research Chemicals
Warning

62470-100G 62470-500G
BJ62470-100GEA 91.87 USD
BJ62470-100G BJ62470-500G
Lithium carbonate ≥99.0% (by titrimetric analysis), Puriss. p.a. ACS for microscopy, Fluka™
Lithium carbonate
Formula: Li₂CO₃
Boiling Pt: 1.300 °C
Melting Pt: 720 °C
Density: 2.110 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011084
CAS Number: 554-13-2
EINECS: 209-062-5

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Insoluble in HCl Max. 0.01 %
Calcium (Ca) Max. 50 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 10 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Heavy metals (ICP) ACS Max. 0.002 %
S-compounds (as SO₄) Max. 50 ppm
Chloride (Cl) Max. 20 ppm
Ammonium (NH₄) Max. 5 ppm
Nitrate (NO₃) Max. 5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR