Lead(II) carbonate basic ≥77% (Pb basis), Puriss., Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Lead carbonat basic, White lead, Lead (II) carbonate hydroxide, Lead (II) hydroxide carbonate

11513-6X1KG 11513-50KG
BJ11513-1KGCS 759 USD
BJ11513-1KG BJ11513-50KG
Lead(II) carbonate basic ≥77% (Pb basis), Puriss., Fluka™
Lead(II) carbonate basic
Formula: (PbCO₃)₂·Pb(OH)₂
MW: 775.63
Melting Pt: 400
Density: 7.06 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00078155
CAS Number: 1319-46-6
EINECS: 215-290-6

Order Now

Specification Test Results

Assay of Pb Min. 77 %
In water soluble matter Max. 0.5 %
Iron (Fe) Max. 0.005 %
Copper (Cu) Max. 0.005 %
Zinc (Zn) Max. 0.005 %
Chloride (Cl) Max. 0.03 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR