Iron(III) sulphate hydrate 21-23% (Fe basis), Puriss. p.a., Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Ferric Persulphate, Diiron trissulphate hydrate, Ferric sulphate hydrate

31235-500G 31235-1KG 31235-6X1KG
BJ31235-500GEA 293.15 USD
BJ31235-500G BJ31235-1KG BJ31235-6X1KG
Iron(III) sulphate hydrate 21-23% (Fe basis), Puriss. p.a., Fluka™
Iron(III) sulphate hydrate

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Formula: Fe₂(SO₄)₃·xH₂O
MW: 399.88 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149714
CAS Number: 15244-10-7
UN: 3260
ADR: 8 III
Merck Index: 13,04065

Order Now

Specification Test Results

Assay of Fe 21 - 23 %
Insoluble in HCl Max. 0.01 %
Arsenic (As) Max. 0.0005 %
Calcium (Ca) Max. 0.01 %
Copper (Cu) Max. 0.002 %
Iron (Fe 2⁺) Max. 0.02 %
Potassium (K) Max. 0.01 %
Magnesium (Mg) Max. 0.01 %
Sodium (Na) Max. 0.01 %
Zinc (Zn) Max. 0.005 %
Chloride (Cl) Max. 0.005 %
Nitrate (NO₃) Max. 0.02 %
Phosphate (PO₄) Max. 0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR