Iron(III) oxide ≥95%, purified, Fluka™
Supplier: Honeywell Research Chemicals
Synonyms:
Ferric oxide
, Diiron trioxide
, Ferric Oxide, (iron(III) oxide)
12342-6X250G
12342-6X1KG
BJ12342-250GCS
290.53
USD
BJ12342-250G
BJ12342-1KG
Iron(III) oxide ≥95%, purified, Fluka™
Iron (III) oxide
Formula:
Fe₂O₃ MW: 159.69 g/mol Melting Pt: 1.565 °C Density: 5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011008 CAS Number: 1309-37-1 EINECS: 215-168-2 |
Specification Test Results
Assay | Min. 95 % |
Loss on ignition | Max. 0.6 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...