Iron(III) chloride hexahydrate ≥99%, Puriss. p.a. Reag. Ph. Eur., Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Iron trichloride hexahydrate, Ferric chloride hexahydrate
31232-6X1KG
31232-250G
31232-25KG
BJ31232-1KGCS
1582.55
USD
BJ31232-1KG
BJ31232-250G
BJ31232-25KG
Iron(III) chloride hexahydrate ≥99%, Puriss. p.a. Reag. Ph. Eur., Fluka™
Iron(III) chloride hexahydrate
Formula:
FeCl₃·6H₂O MW: 270.3 g/mol Boiling Pt: 280…285 °C Melting Pt: 37 Density: 2.8 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149712 CAS Number: 10025-77-1 UN: 3260 ADR: 8 III Merck Index: 13,04048 |
|
Specification Test Results
Assay | Min. 99 % |
Free acid (as HCl) | Max. 0.2 % |
Arsenic (As) | Max. 0.0005 % |
Copper (Cu) | Max. 0.002 % |
Iron (Fe 2⁺) | Max. 0.005 % |
Lead (Pb) | Max. 0.002 % |
Zinc (Zn) | Max. 0.002 % |
Substances not precipitated by NH₄OH (SO₄) | Max. 0.05 % |
Free chlorine (Cl) | Max. 0.001 % |
Phosphate (PO₄) | Max. 0.002 % |
Sulfate (SO₄) | Max. 0.005 % |
Total N | Max. 0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...