Glycerine ≥99.0% (by GC), Reagent Grade

Supplier: Honeywell Research Chemicals

Synonyms: 1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol

G7757-1GA G7757-1L G7757-20L G7757-5L
BJG7757-1GAEA 276 USD
BJG7757-1GA BJG7757-1L BJG7757-20L BJG7757-500ML BJG7757-5L
Glycerine ≥99.0% (by GC), Reagent Grade
Glycerine
Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290
Melting Pt: 18.6
Density: 1.26 g/cm³ (20 °C)
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Vapor Density 3.1 (vs air)
Identity (IR) complying
Assay (GC) Min. 99.0 %
Water (Karl Fischer) Max. 1.0 %
Sulfated ash Max. 0.1 %
Heavy metals (as Pb) Max. 5 ppm
Vapor Pressure <1 mmHg ( 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR