Copper(II) carbonate basic ≥95% (RT), Purum p.a.
Supplier: Honeywell Research Chemicals
|
Warning
|
Synonyms:
Copper (II) carbonate dihydroxide, Copper(II) carbonate, basic, Basic copper carbonate, Copper (II) carbonate-copper (II) hydroxide (1:1)
61167-1KG
61167-250G
BJ61167-1KGEA
284.99
USD
BJ61167-1KG
BJ61167-250G
Copper(II) carbonate basic ≥95% (RT), Purum p.a.
Copper(II) carbonate basic
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Formula:
CuCO₃·Cu(OH)₂ MW: 221.12 g/mol Melting Pt: 270 °C Density: 4 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00010976 CAS Number: 12069-69-1 EINECS: 235-113-6 |
Specification Test Results
Assay | 95.0 - 105.0 % |
Calcium (Ca) | Max. 5000 ppm |
Cadmium (Cd) | Max. 50 ppm |
Cobalt (Co) | Max. 50 ppm |
Iron (Fe) | Max. 100 ppm |
Potassium (K) | Max. 100 ppm |
Sodium (Na) | Max. 5000 ppm |
Nickel (Ni) | Max. 50 ppm |
Lead (Pb) | Max. 50 ppm |
Zinc (Zn) | Max. 1000 ppm |
Total S (as SO₄) | Max. 200 ppm |
Chloride (Cl) | Max. 1000 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...