Cobalt(II) nitrate hexahydrate ≥98% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Cobalt dinitrate hexahydrate, Cobaltous nitrate hexahydrate

239267-500G 239267-5G
BJ239267-100GEA 107.81 USD
BJ239267-100G BJ239267-500G BJ239267-5G
Cobalt(II) nitrate hexahydrate ≥98% ACS, Fluka™
Cobalt(II) nitrate hexahydrate
Formula: Co(NO₃)₂·6H₂O
MW: 291.03 g/mol
Boiling Pt: Dec. (on heating)
Melting Pt: 55…56 °C
Density: 1.87 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149647
CAS Number: 10026-22-9
UN: 1477
ADR: 5.1,II

Order Now

Specification Test Results

Appearance of the substance complying
Assay 98.0 - 102.0 %
Water insoluble matter Max. 0.01 %
Calcium (Ca) Max. 0.005 %
Copper (Cu) Max. 0.002 %
Iron (Fe) Max. 0.001 %
Potassium (K) Max. 0.01 %
Magnesium (Mg) Max. 0.005 %
Sodium (Na) Max. 0.05 %
Nickel (Ni) Max. 0.15 %
Lead (Pb) Max. 0.002 %
Zinc (Zn) Max. 0.01 %
Chloride (Cl) Max. 0.002 %
Sulfate (SO₄) Max. 0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR