Calcium sulphate hemihydrate ≥97.0%, Purum, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Plaster of Paris

12090-1KG 12090-25KG
BJ12090-1KGEA 85 USD
BJ12090-1KG BJ12090-25KG
Calcium sulphate hemihydrate ≥97.0%, Purum, Fluka™
Calcium sulphate hemihydrate
Formula: CaSO₄·0.5H₂O
MW: 145.15
Density: 2.320 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010912
CAS Number: 10034-76-1

Order Now

Specification Test Results

Assay Min. 97 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR