Calcium nitrate tetrahydrate 99-103%, Puriss. p.a. ACS, Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
31218-1KG
31218-500G
31218-5KG
BJ31218-1KGEA
193.65
USD
BJ31218-1KG
BJ31218-250G
BJ31218-500G
BJ31218-5KG
Calcium nitrate tetrahydrate 99-103%, Puriss. p.a. ACS, Fluka™
Calcium nitrate tetrahydrate
Formula:
Ca(NO₃)₂·4H₂O MW: 236.15 g/mol Melting Pt: 42 °C Density: 1.820 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149604 CAS Number: 13477-34-4 |
Specification Test Results
Assay | 99.0 - 103.0 % |
pH (5 %, 25 °C) | 5.0 - 7.0 |
Water insoluble matter | Max. 0.005 % |
Barium (Ba) | Max. 0.005 % |
Iron (Fe) | Max. 5 ppm |
Potassium (K) | Max. 0.005 % |
Magnesium (Mg) | Max. 0.01 % |
Sodium (Na) | Max. 0.01 % |
Strontium (Sr) | Max. 0.01 % |
Heavy metals (ICP) ACS | Max. 5 ppm |
Ammonium (NH₄) | Max. 0.005 % |
Chloride (Cl) | Max. 0.002 % |
Nitrite (NO₂) | Max. 0.001 % |
Phosphate (PO₄) | Max. 0.001 % |
Sulfate (SO₄) | max. 0.002 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...