Calcium hydroxide ≥96%, Puriss. p.a. Reag. Ph. Eur., Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
31219-100G
31219-6X1KG
31219-25KG
31219-500G
BJ31219-100GEA
68.18
USD
BJ31219-100G
BJ31219-1KG
BJ31219-25KG
BJ31219-500G
Calcium hydroxide ≥96%, Puriss. p.a. Reag. Ph. Eur., Fluka™
Calcium hydroxide
Formula:
Ca(OH)₂ Boiling Pt: Decomposes on heating Melting Pt: 580 °C Density: 2.24 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010901 CAS Number: 1305-62-0 EINECS: 215-137-3 |
Specification Test Results
Assay | Min. 96 % |
Assay of CaCO₃ | Max. 3 % |
Insoluble in hydrochloric acid | Max. 0.03 % |
Copper (Cu) | Max. 0.0005 % |
Iron (Fe) | Max. 0.05 % |
Lead (Pb) | Max. 0.0002 % |
Zinc (Zn) | Max. 0.0005 % |
Substances not precipitated by NH₄OH (asO₄) | Max. 2.5 % |
Chloride (Cl) | Max. 0.005 % |
Sulfate (SO₄) | Max. 0.05 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...