Calcium chloride dihydrate ≥99%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Warning

31307-1KG 31307-6X2.5KG
BJ31307-1KGEA 316.42 USD
BJ31307-1KG BJ31307-2.5KG BJ31307-500G
Calcium chloride dihydrate ≥99%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Calcium chloride dihydrate
Formula: CaCl₂·2H₂O
Boiling Pt: 1.900 °C (1.013 hPa)
Melting Pt: 765 °C
Storage Temperature: Ambient
MDL Number: MFCD00149613
CAS Number: 10035-04-8
EINECS: 233-140-8 

Order Now

Specification Test Results

Assay 99 - 105.0 %
pH (5 %, 25 °C) 4.5 - 8.0
Water insoluble matter Max. 0.01 %
Acidity or alkalinity complying
Aluminium (Al) Max. 0.0001 %
Arsenic (As) Max. 1 ppm
Barium (Ba) Max. 0.005 %
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 0.005 %
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 0.0005 %
Potassium (K) Max. 0.005 %
Magnesium (Mg) Max. 0.005 %
Sodium (Na) Max. 0.005 %
Nickel (Ni) Max. 0.005 %
Lead (Pb) Max. 5 ppm
Strontium (Sr) Max. 0.01 %
Zinc (Zn) Max. 0.0005 %
Mg and alkali salts (SO₄) Max. 0.5 %
Heavy metals (ICP) ACS Max. 5 ppm
Ammonium (NH₄) Max. 0.005 %
Nitrate (NO₃) Max. 0.002 %
Sulfate (SO₄) Max. 0.005 %
Oxidizing substances (as NO₃) Max. 0.003 %
Appearance of the solution complying
Colour of the solution complying
Turbidity Max. 3.5 NTU

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR