Bromophenol blue Reag. Ph. Eur. indicator, Fluka™
Supplier: Honeywell Research Chemicals
Warning
|
Synonyms:
Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
32712-25G
32712-5G
BJ32712-25GEA
213.09
USD
BJ32712-25G
BJ32712-5G
Bromophenol blue Reag. Ph. Eur. indicator, Fluka™
Bromophenol blue
Formula:
C₁₉H₁₀Br₄O₅S MW: 669.97 g/mol Melting Pt: 204 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005875 CAS Number: 115-39-9 EINECS: 204-086-2 |
Specification Test Results
Indicator solution | complying |
Test of sensitivity | complying |
Change range (pH 2,8-4,4) | complying |
UV spectrum | complying |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...