Bromocresol purple Reag. Ph. Eur. indicator, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: 5',5''-Dibromo-o-cresolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

32642-1KG 32642-25G 32642-5G
BJ32642-1KGEA 2402.77 USD
BJ32642-1KG BJ32642-25G BJ32642-5G
Bromocresol purple Reag. Ph. Eur. indicator, Fluka™
Bromocresol purple
Formula: C₂₁H₁₆Br₂O₅S
MW: 540.23 g/mol
Melting Pt: 240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011681
CAS Number: 115-40-2
EINECS: 204-087-8

Order Now

Specification Test Results

Change range complying
Indicator solution complying
Test of sensitivity complying
Curve course (UV) complying
Extinction maximum at 414 - 424 nm
Extinction maximum of Min. 400
pH Range 5.2 - 6.8, yellow to purple

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR