Barium hydroxide octahydrate ≥98%, Puriss. p.a. ISO, Reag. Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Danger

31127-100G 31127-1KG
BJ31127-100GEA 78.91 USD
BJ31127-100G BJ31127-1KG
Barium hydroxide octahydrate ≥98%, Puriss. p.a. ISO, Reag. Ph. Eur., Fluka™
Barium hydroxide octahydrate
Formula: Ba(OH)₂·8H₂O
Boiling Pt: Dec. (on heating)
Melting Pt: 78 °C
Density: 2.180 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149152
CAS Number: 12230-71-6
EINECS: 241-234-5
UN: 3262
ADR: 8,II

Order Now

Specification Test Results

Assay Min. 98 %
Assay of BaCO₃ Max. 2 %
Insoluble in acid Max. 0.005 %
Calcium (Ca) Max. 0.005 %
Cadmium (Cd) Max. 0.0005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 0.0005 %
Potassium (K) Max. 0.005 %
Magnesium (Mg) Max. 0.002 %
Sodium (Na) Max. 0.005 %
Lead (Pb) Max. 0.0005 %
Strontium (Sr) Max. 1.5 %
Zinc (Zn) Max. 0.0005 %
Heavy metals (as Pb) Max. 0.0005 %
Chloride (Cl) Max. 0.001 %
Sulfide (S) Max. 0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR