Arsenazo III ≥79% for spectrophotometric determination of Th, Zr, U, Cd, Zn, Ca, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: 2,7-Bis(2-arsonophenylazo)chromotropic acid, 2,7-Bis(2-arsonophenylazo)-1,8-dihydroxynaphthalene-3,6-disulphonic acid, 2,2′-(1,8-Dihydroxy-3,6-disulphonaphthylene-2,7-bisazo)bisbenzenearsonic acid, Arsenazo Ⅲ

11090-25G 11090-5G
BJ11090-25GEA 1030.24 USD
BJ11090-25G BJ11090-5G
Arsenazo III ≥79% for spectrophotometric determination of Th, Zr, U, Cd, Zn, Ca, Fluka™
Arsenazo III
Formula: C₂₂H₁₈As₂N₄O₁₄S₂
MW: 776.38 g/mol
Boiling Pt: Decomposition
Melting Pt: 320 °C
Storage Temperature: Ambient
MDL Number: MFCD00036695
CAS Number: 1668-00-4
EINECS: 216-788-6
UN: 3280
ADR: 6.1,II

Order Now

Specification Test Results

Assay (HPLC) Min. 79 %
Colour of the solution complying
Loss on drying (110°C) Max. 10 %
Sulfated ash Max. 1 %
Calcium (Ca) Max. 500 ppm
Cadmium (Cd) Max. 50 ppm
Chromium (Cr) Max. 200 ppm
Magnesium (Mg) Max. 50 ppm
Molybdenum (Mo) Max. 50 ppm
Zinc (Zn) Max. 50 ppm
Absorbtion (1%/1cm) Min. 300
Absorption maximum 535 - 543 nm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR