Ammonium trifluoroacetate ≥99.0% (by non aqueous titration) for HPLC, Fluka™
Supplier: Honeywell Research Chemicals
Synonyms:
Ammoniumtrifluoroacetate, Trifluoroacetic acid ammonium salt
17839-10G
17839-50G
BJ17839-10GEA
206
USD
BJ17839-10G
BJ17839-50G
Ammonium trifluoroacetate ≥99.0% (by non aqueous titration) for HPLC, Fluka™
Ammonium trifluoroacetate
Formula:
C₂H₄F₃NO₂ Melting Pt: 123…125 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012615 CAS Number: 3336-58-1 EINECS: 222-072-4 |
Specification Test Results
Assay | 99.0 - 101.0 % |
Identity (IR) | complying |
pH (10 %, 20°C) | 5 - 6.5 |
Colour of the solution | complying |
Turbidity of solution | complying |
Suitability test (HPLC) | complying |
Absorbance at 500 nm | Max. 0.02 |
Absorbance at 290 nm | Max. 0.02 |
Absorbance at 280 nm | Max. 0.03 |
Absorbance at 270 nm | Max. 0.04 |
Absorbance at 260 nm | Max. 0.05 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...