Ammonium fluoride ≥98%, Puriss. p.a. ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Fluoroammonium

30101-100G 30101-6X1KG
BJ30101-100GEA 108 USD
BJ30101-100G BJ30101-1KG
Ammonium fluoride ≥98%, Puriss. p.a. ACS, Fluka™
Ammonium fluoride
Formula: NH₄F
MW: 37.04 g/mol
Melting Pt: 100 °C
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011423
CAS Number: 12125-01-8
EINECS: 235-185-9
UN: 2505
ADR: 6.1,III

Order Now

Specification Test Results

Assay Min. 98 %
Ammonium hydrogen difluoride (NH₄HF₂) Max. 1 %
Water insoluble matter Max. 0.005 %
Residue on ignition (SO₄) Max. 0.005 %
Cadmium (Cd) Max. 0.0005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 0.0005 %
Potassium (K) Max. 0.002 %
Sodium (Na) Max. 0.002 %
Lead (Pb) Max. 0.0005 %
Zinc (Zn) Max. 0.0005 %
Heavy metals (as Pb) Max. 0.0005 %
Chloride (Cl) Max. 0.0005 %
Hexafluorosilicicate (as SiF₆) Max. 0.1 %
Sulfate (SO₄) Max. 0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR