Ammonium cerium(IV) sulphate dihydrate ≥98.0% (RT), Puriss. p.a. ACS, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Tetraammonium cerium tetrakis(sulphate) dihydrate, Cerium (IV) ammonium sulphate dihydrate, Tetraammonium cerium(IV) sulphate dihydrate, Ammonium cerium(IV) sulphate hydrate, Ammonium ceric sulphate dihydrate

22269-100G 22269-500G
BJ22269-100GEA 242.47 USD
BJ22269-100G BJ22269-500G
Ammonium cerium(IV) sulphate dihydrate ≥98.0% (RT), Puriss. p.a. ACS, Fluka™
Ammonium cerium(IV) sulphate dihydrate
Formula: (NH₄)₂Ce(SO₄)₄·2H₂O
MW: 632.55 g/mol
Melting Pt: 130
Storage Temperature: Ambient
MDL Number: MFCD00149417
CAS Number: 10378-47-9

Order Now

Specification Test Results

Assay (Iodometric) 98.0 - 102.0 %
Insoluble in acid Max. 0.05 %
Iron (Fe) Max. 100 ppm
Phosphate (PO₄) Max. 300 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR