Amidosulfonic acid ≥99.5% (by alkalimetry)
Supplier: Honeywell Research Chemicals
|
Warning
|
Synonyms:
Aminosulphonic acid, Amidosulphuric acid, Amidosulphuric Acid, Sulphamic acid, Amidosulphonic acid, Sulphamidic acid
07401-1KG
07401-6X2.5KG
BJ07401-1KGEA
63
USD
BJ07401-1KG
BJ07401-2.5KG
BJ07401-5KG
Amidosulfonic acid ≥99.5% (by alkalimetry)
Amidosulphonic acid
Formula:
H₃NO₃S MW: 97.09 g/mol Melting Pt: 209 °C.205 °C Density: 2.1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011603 CAS Number: 5329-14-6 EINECS: 226-218-8 UN: 2967 ADR: 8,III |
Specification Test Results
Assay (alkalimetric) | Min. 99.5 % |
Residue on ignition | Max. 0.02 % |
Iron (Fe) | Max. 0.003 % |
Heavy metals (as Pb) | Max. 0.001 % |
Chloride (Cl) | Max. 0.001 % |
Sulfate (SO₄) | Max. 0.15 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...