Aluminium ammonium sulphate dodecahydrate ≥99% (by titrimetric analysis), Reagent Grade

Supplier: Honeywell Research Chemicals

Synonyms: Aluminium ammonium bis(sulphate) dodecahydrate, Ammonium alum, Ammonium aluminum sulphate dodecahydrate, Aluminum ammonium sulphatedodecahydrate

A2140-1KG A2140-500G
BJA2140-1KGEA 314.1 USD
BJA2140-1KG BJA2140-500G
Aluminium ammonium sulphate dodecahydrate ≥99% (by titrimetric analysis), Reagent Grade
Aluminium ammonium sulphate dodecahydrate
Formula: AlNH₄(SO₄)₂·12H₂O
MW: 453.33 g/mol
Boiling Pt: 200
Melting Pt: 93
Density: 1.640 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00051042
CAS Number: 7784-26-1
EINECS: 232-055-3
Merck Index: 13,00325

Order Now

Specification Test Results

Assay Min. 99 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR